ENA432539930 Enamine

2-bromo-1-(4-propylphenyl)ethan-1-one

  • MDL number MFCD11131363
  • Cas Number : 64328-67-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-197226

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197226
Availability details for
Quantity
  • Availability
  • Add to Cart