ENA432539931 Enamine

5-bromo-2-(1-chloroethyl)pyrimidine

  • MDL number MFCD23958253
  • Cas Number : 1514124-47-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-197227

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197227
Availability details for
Quantity
  • Availability
  • Add to Cart