ENA432539936 Enamine

tert-butyl 3-(aminomethyl)-3-hydroxyazetidine-1-carboxylate

  • MDL number MFCD17015986
  • Cas Number : 1008526-71-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-197235

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197235
Availability details for
Quantity
  • Availability
  • Add to Cart