ENA432539939 Enamine

methyl 2-(3-bromo-2-nitrophenyl)acetate

  • MDL number MFCD18398873
  • Cas Number : 1261653-58-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-197259

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197259
Availability details for
Quantity
  • Availability
  • Add to Cart