ENA432539945 Enamine

5-chloro-6-nitro-2,3-dihydro-1,3-benzoxazol-2-one

  • MDL number MFCD00598829
  • Cas Number : 27087-06-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-197267

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197267
Availability details for
Quantity
  • Availability
  • Add to Cart