ENA432539946 Enamine

[2-(diethylamino)pyrimidin-5-yl]boronic acid

  • MDL number MFCD07375163
  • Cas Number : 1888409-34-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-197268

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197268
Availability details for
Quantity
  • Availability
  • Add to Cart