ENA432539947 Enamine

N-propyl-8-(trifluoromethoxy)quinolin-4-amine

  • MDL number MFCD17312572
  • Cas Number : 1275172-96-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195439

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195439
Availability details for
Quantity
  • Availability
  • Add to Cart