ENA432539948 Enamine

9-fluoro-1H,2H,3H,4H,5H,10H-azepino[3,4-b]indole

  • MDL number MFCD11168335
  • Cas Number : 1042626-47-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-197270

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197270
Availability details for
Quantity
  • Availability
  • Add to Cart