ENA432539953 Enamine

3-ethyl-1-methyl-1,4-diazepane-2,5-dione

  • MDL number MFCD20434939
  • Cas Number : 1483527-25-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-195441

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195441
Availability details for
Quantity
  • Availability
  • Add to Cart