ENA432539954 Enamine

hex-5-en-3-amine hydrochloride

  • MDL number MFCD08451503
  • Cas Number : 852879-02-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-197280

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197280
Availability details for
Quantity
  • Availability
  • Add to Cart