ENA432539957 Enamine

2-hydroxy-3,5-dimethylbenzene-1-sulfonyl chloride

  • MDL number MFCD19200784
  • Cas Number : 872277-84-4
  • Molecular Weight : 220.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-hydroxy-3,5-dimethylbenzenesulfonyl chloride
Inchi Code 1S/C8H9ClO3S/c1-5-3-6(2)8(10)7(4-5)13(9,11)12/h3-4,10H,1-2H3
Inchi Key VDCHDSXNEUWUGQ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-197291

Safety Information

signalword Danger
pcodes P260,P261,P264,P270,P271,P280,P301+P312,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P330,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-197291
pictograms GHS05,GHS07
hstatements H302,H314,H335
Availability details for
Quantity
  • Availability
  • Add to Cart