ENA432539961 Enamine

5,5-dimethyloxolane-2-carbaldehyde

  • MDL number MFCD24677374
  • Cas Number : 144640-92-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-197327

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197327
Availability details for
Quantity
  • Availability
  • Add to Cart