ENA432539962 Enamine

2-methylindolizine-6-carboxylic acid

  • MDL number MFCD08056317
  • Cas Number : 256935-78-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-197331

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197331
Availability details for
Quantity
  • Availability
  • Add to Cart