ENA432539981 Enamine

2-methyl-4-(pyrrolidin-1-yl)butan-2-ol

  • MDL number MFCD21858978
  • Cas Number : 1512285-97-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-197392

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197392
Availability details for
Quantity
  • Availability
  • Add to Cart