ENA432539984 Enamine

1-(4-hydroxycyclohexyl)pyrrolidin-2-one

  • MDL number MFCD26905212
  • Cas Number : 895581-20-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-197398

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197398
Availability details for
Quantity
  • Availability
  • Add to Cart