ENA432539987 Enamine

2-bromo-1-cyclopropylpropan-1-one

  • MDL number MFCD13172605
  • Cas Number : 34650-66-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-197404

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197404
Availability details for
Quantity
  • Availability
  • Add to Cart