ENA432539989 Enamine

5-(pyrimidin-5-yl)pyridin-2-amine

  • MDL number MFCD06802737
  • Cas Number : 827588-84-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-197407

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197407
Availability details for
Quantity
  • Availability
  • Add to Cart