ENA432539990 Enamine

N-tert-butyl-1-chloromethanesulfonamide

  • MDL number MFCD09865763
  • Cas Number : 115719-94-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-197410

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197410
Availability details for
Quantity
  • Availability
  • Add to Cart