ENA432539991 Enamine

7,7-dimethyl-1,3-diazaspiro[4.4]nonane-2,4-dione

  • MDL number MFCD23969019
  • Cas Number : 1536184-51-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-197416

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197416
Availability details for
Quantity
  • Availability
  • Add to Cart