ENA432540004 Enamine

2-(1,1-dioxo-2,3-dihydro-1lambda6-benzothiophen-3-yl)acetic acid

  • MDL number MFCD12190910
  • Cas Number : 130105-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-195456

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195456
Availability details for
Quantity
  • Availability
  • Add to Cart