ENA432540009 Enamine

4-(4-chlorophenyl)-4-fluoropiperidine hydrochloride

  • MDL number MFCD28145513
  • Cas Number : 324002-51-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-197471

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197471
Availability details for
Quantity
  • Availability
  • Add to Cart