ENA432540013 Enamine

1,1,1-trifluoro-3-(pyridin-4-yl)propan-2-ol

  • MDL number MFCD16617632
  • Cas Number : 1491591-94-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-197514

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197514
Availability details for
Quantity
  • Availability
  • Add to Cart