ENA432540018 Enamine

[3-(hydroxymethyl)phenyl]methanesulfonamide

  • MDL number MFCD21298917
  • Cas Number : 1493996-36-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-197540

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197540
Availability details for
Quantity
  • Availability
  • Add to Cart