ENA432540022 Enamine

1-phenylpent-4-en-2-ol

  • MDL number MFCD00598882
  • Cas Number : 61077-65-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-197556

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197556
Availability details for
Quantity
  • Availability
  • Add to Cart