ENA432540027 Enamine

3-(cyclopentanesulfonyl)-2,6-difluorobenzoic acid

  • MDL number MFCD12633229
  • Cas Number : 1097007-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-195459

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195459
Availability details for
Quantity
  • Availability
  • Add to Cart