ENA432540028 Enamine

4-[(4-amino-3-fluorophenyl)methyl]-1-methylpiperazin-2-one

  • MDL number MFCD22920912
  • Cas Number : 1539447-43-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195460

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195460
Availability details for
Quantity
  • Availability
  • Add to Cart