ENA432540029 Enamine

[2-(4-amino-3-methylphenoxy)acetyl]urea

  • MDL number MFCD09044623
  • Cas Number : 926269-03-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-195461

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195461
Availability details for
Quantity
  • Availability
  • Add to Cart