ENA432540031 Enamine

[2-(2-tert-butylphenoxy)ethyl](methyl)amine hydrochloride

  • MDL number MFCD28139616
  • Cas Number : 1628167-50-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-195462

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195462
Availability details for
Quantity
  • Availability
  • Add to Cart