ENA432540043 Enamine

6-bromo-3,5-dimethylimidazo[1,2-a]pyridine

  • MDL number MFCD24199655
  • Cas Number : 1545482-60-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-197642

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197642
Availability details for
Quantity
  • Availability
  • Add to Cart