ENA432540044 Enamine

prop-2-en-1-yl 2-hydroxypropanoate

  • MDL number MFCD00046637
  • Cas Number : 5349-55-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-197644

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197644
Availability details for
Quantity
  • Availability
  • Add to Cart