ENA432540045 Enamine

6,6-dimethyl-1,3-diazaspiro[4.4]nonane-2,4-dione

  • MDL number MFCD20729135
  • Cas Number : 90565-74-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-197645
Availability details for
Quantity
  • Availability
  • Add to Cart