ENA432540046 Enamine

3-(3,4-dichlorophenyl)-2-formamidopropanoic acid

  • MDL number MFCD19661063
  • Cas Number : 52715-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-197647

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197647
Availability details for
Quantity
  • Availability
  • Add to Cart