ENA432540049 Enamine

3-(1H-1,3-benzodiazol-2-yl)-2,2-dimethylpropanoic acid

  • MDL number MFCD05625042
  • Cas Number : 95242-09-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-197650

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197650
Availability details for
Quantity
  • Availability
  • Add to Cart