ENA432540055 Enamine

5-(aminomethyl)-2-methylfuran-3-carboxylic acid hydrochloride

  • MDL number MFCD28145525
  • Cas Number : 1803592-68-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-197676

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197676
Availability details for
Quantity
  • Availability
  • Add to Cart