ENA432540056 Enamine

2-chloro-4-fluoro-1-methyl-1H-1,3-benzodiazole

  • MDL number MFCD17168976
  • Cas Number : 1283719-98-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-197678

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197678
Availability details for
Quantity
  • Availability
  • Add to Cart