ENA432540059 Enamine

methyl 2-{[2-(4-methylpiperazin-1-yl)ethyl]amino}acetate

  • MDL number MFCD24325229
  • Cas Number : 1553196-16-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-197685

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197685
Availability details for
Quantity
  • Availability
  • Add to Cart