ENA432540063 Enamine

4-(4-bromobutyl)oxane

  • MDL number MFCD20343085
  • Cas Number : 1050496-76-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-197737

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197737
Availability details for
Quantity
  • Availability
  • Add to Cart