ENA432540064 Enamine

1-(4-chlorophenyl)-4-methyl-1H-pyrazol-3-amine

  • MDL number MFCD16845434
  • Cas Number : 1247390-15-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-197772

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197772
Availability details for
Quantity
  • Availability
  • Add to Cart