ENA432540071 Enamine

2-amino-2-(3-chloro-2-fluorophenyl)ethan-1-ol hydrochloride

  • MDL number MFCD28145531
  • Cas Number : 1245623-52-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-197821

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197821
Availability details for
Quantity
  • Availability
  • Add to Cart