ENA432540076 Enamine

methyl 4-bromo-2-fluoro-5-iodobenzoate

  • MDL number MFCD24107121
  • Cas Number : 1509454-55-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-194976

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194976
Availability details for
Quantity
  • Availability
  • Add to Cart