ENA432540083 Enamine

1,3-diethyl 2-(4-bromophenyl)-4-hydroxy-4-methyl-6-oxocyclohexane-1,3-dicarboxylate

  • MDL number MFCD00521058
  • Cas Number : 4759-57-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-197887

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197887
Availability details for
Quantity
  • Availability
  • Add to Cart