ENA432540086 Enamine

5-(2-aminoethyl)pyridin-3-ol

  • MDL number MFCD22412542
  • Cas Number : 1803585-03-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-197919

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197919
Availability details for
Quantity
  • Availability
  • Add to Cart