ENA432540087 Enamine

4-fluoro-2-methylphenyl chloroformate

  • MDL number MFCD24237682
  • Cas Number : 1552201-86-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195494

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195494
Availability details for
Quantity
  • Availability
  • Add to Cart