ENA432540182 Enamine

2-(4-methoxycyclohexyl)-2-[(trimethylsilyl)oxy]propanenitrile

  • MDL number MFCD28145549
  • Cas Number : 1955561-94-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-198086

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198086
Availability details for
Quantity
  • Availability
  • Add to Cart