ENA432540187 Enamine

4-(methoxymethoxy)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

  • MDL number MFCD28145560

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-198697

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198697
Availability details for
Quantity
  • Availability
  • Add to Cart