ENA432540216 Enamine

(2R)-2-amino-N-[2-(piperidin-1-yl)ethyl]propanamide dihydrochloride

  • MDL number MFCD28145596
  • Cas Number : 1807938-17-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-199088

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199088
Availability details for
Quantity
  • Availability
  • Add to Cart