ENA432540218 Enamine

3-[6-(3-hydroxyprop-1-yn-1-yl)-2-methylpyrimidin-4-yl]prop-2-yn-1-ol

  • MDL number MFCD28145599
  • Cas Number : 1803572-31-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-199100

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199100
Availability details for
Quantity
  • Availability
  • Add to Cart