ENA432540225 Enamine

1-amino-4,4-dimethyl-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid hydrochloride

  • MDL number MFCD28145611
  • Cas Number : 1803567-18-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-199166

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199166
Availability details for
Quantity
  • Availability
  • Add to Cart