ENA432540240 Enamine

[(4-chlorophenyl)methyl](3-methylbut-2-en-1-yl)amine hydrochloride

  • MDL number MFCD28145627
  • Cas Number : 1803560-81-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-199312

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199312
Availability details for
Quantity
  • Availability
  • Add to Cart