ENA432540247 Enamine

(2-methoxyethyl)[1-(1,3-thiazol-2-yl)ethyl]amine hydrochloride

  • MDL number MFCD28145637
  • Cas Number : 1803584-16-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-199425

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199425
Availability details for
Quantity
  • Availability
  • Add to Cart